6-Hydroxy-7-methoxyquinazolin-4(1H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxy-7-methoxy-4(3H)-quinazolinone, 6-Hydroxy-7-methoxy-3H-quinazolin-4-one

A230420-100G A230420-10G A230420-1G A230420-25G A230420-5G
77000-164EA 89 USD
77000-164 77000-166 77000-168 77000-170 77000-172
6-Hydroxy-7-methoxyquinazolin-4(1H)-one 98%
6-Hydroxy-7-methoxyquinazolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O₃
MDL Number: MFCD20547768
CAS Number: 179688-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR