8-Methoxy-2-(trifluoromethyl)quinoline-5-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A230395-100MG
A230395-1G
A230395-250MG
A230395-5G
76987-326EA
111.4
USD
76987-326
76987-328
76987-330
76987-332
8-Methoxy-2-(trifluoromethyl)quinoline-5-carboxylic acid 95%
8-Methoxy-2-(trifluoromethyl)quinoline-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₂H₈F₃NO₃ MW: 271.19 g/mol |
MDL Number:
MFCD20272020 CAS Number: 199872-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...