Methyl-4-tert-butylbenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(tert-butyl)benzoate

A230363-100G A230363-10G A230363-25G A230363-500G
77000-156EA 15.92 USD
77000-156 77000-158 77000-160 77000-162
Methyl-4-tert-butylbenzoate 95%
Methyl 4-tert-butylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
MDL Number: MFCD00008835
CAS Number: 26537-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR