2,6-Dibromo-4,4-dihexyl-4H-cyclopenta[1,2-b:5,4-b']dithiophene 97%
Supplier: Ambeed
|
Warning
|
A230318-100MG
A230318-1G
A230318-250MG
76987-320EA
53
USD
76987-320
76987-322
76987-324
2,6-Dibromo-4,4-dihexyl-4H-cyclopenta[1,2-b:5,4-b']dithiophene 97%
2,6-Dibromo-4,4-dihexyl-4H-cyclopenta[1,2-b:5,4-b']dithiophene
Precaución: for Research Use Only
Formula:
C₂₁H₂₈Br₂S₂ |
MDL Number:
MFCD16619321 CAS Number: 528570-55-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...