5-Acetyl-2-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (5-Acetyl-2-methoxyphenyl)boronic acid

A230304-1G A230304-250MG A230304-5G
77031-028EA 23 USD
77031-028 77031-030 77031-032
5-Acetyl-2-methoxyphenylboronic acid 97%
5-Acetyl-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MW: 193.992 g/mol
MDL Number: MFCD09839106
CAS Number: 1215281-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR