4,5-Dichloro-2-fluorobenzoic acid 95%

Supplier: Ambeed
Warning

A230190-1G A230190-5G
77000-154EA 10 USD
77000-154 77881-594
4,5-Dichloro-2-fluorobenzoic acid 95%
4,5-Dichloro-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO₂
MDL Number: MFCD00672988
CAS Number: 289039-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR