Ethyl 1-(5-bromopyridin-3-yl)piperidine-4-carboxylate 97%

Supplier: Ambeed
Warning

A230136-1G A230136-250MG
76987-314EA 140.04 USD
76987-314 76987-316
Ethyl 1-(5-bromopyridin-3-yl)piperidine-4-carboxylate 97%
Ethyl 1-(5-bromopyridin-3-yl)piperidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₇BrN₂O₂
MW: 313.19 g/mol
MDL Number: MFCD14706144
CAS Number: 847406-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR