6-Chloro-2-fluoropurine 98%

Supplier: Ambeed
Warning

A230111-100G A230111-10G A230111-1G A230111-250MG A230111-25G A230111-5G
77031-014EA 315 USD
77031-014 77031-016 77031-018 77031-020 77031-022 77031-024
6-Chloro-2-fluoropurine 98%
6-Chloro-2-fluoropurine

Precaución: for Research Use Only

Formula: C₅H₂ClFN₄
MDL Number: MFCD02183557
CAS Number: 1651-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR