(R)-4-(3-Bromophenyl)-3-((tert-butoxycarbonyl)amino)butanoic acid 98%

Supplier: Ambeed
Warning

A229868-1G A229868-250MG
76987-308EA 163.38 USD
76987-308 76987-310
(R)-4-(3-Bromophenyl)-3-((tert-butoxycarbonyl)amino)butanoic acid 98%
(R)-4-(3-Bromophenyl)-3-((tert-butoxycarbonyl)amino)butanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₀BrNO₄
MW: 358.22 g/mol
MDL Number: MFCD11617947
CAS Number: 1350734-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR