Rabeprazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-(((4-(3-Methoxypropoxy)-3-methylpyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole

A229737-10MG A229737-1MG A229737-50MG A229737-5MG
77031-006EA 36.08 USD
77031-006 77031-008 77031-010 77031-012
Rabeprazole 98%
Rabeprazole

Precaución: for Research Use Only

Formula: C₁₈H₂₁N₃O₃S
MW: 359.45 g/mol
MDL Number: MFCD00868879
CAS Number: 117976-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR