5H-Benzo[d]benzo[4,5]imidazo[1,2-a]imidazole 95%

Supplier: Ambeed
Warning

A229639-1G A229639-250MG
76987-294EA 134 USD
76987-294 76987-296
5H-Benzo[d]benzo[4,5]imidazo[1,2-a]imidazole 95%
5H-Benzo[d]benzo[4,5]imidazo[1,2-a]imidazole

Precaución: for Research Use Only

Formula: C₁₃H₉N₃
MDL Number: MFCD24551866
CAS Number: 28890-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR