1-(3,5-Dimethylfuran-2-yl)ethanone 95%

Supplier: Ambeed
Warning

A229525-100MG A229525-1G A229525-250MG
76987-286EA 179.3 USD
76987-286 76987-288 76987-290
1-(3,5-Dimethylfuran-2-yl)ethanone 95%
1-(3,5-Dimethylfuran-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.16 g/mol
MDL Number: MFCD00052576
CAS Number: 22940-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR