tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazine-4(3H)-carboxylate 97%

Supplier: Ambeed
Warning

A229357-100MG A229357-250MG A229357-1G
76987-276EA 119.89 USD
76987-276 76987-278 77612-232
tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazine-4(3H)-carboxylate 97%
tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[b][1,4]oxazine-4(3H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₉H₂₈BNO₅
MW: 361.24 g/mol
MDL Number: MFCD16036418
CAS Number: 1161362-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR