Methyl-2,4-dichloronicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,4-dichloronicotinate

A229147-1G A229147-250MG A229147-5G A229147-10G A229147-25G
77000-146EA 46.68 USD
77000-146 77000-148 77000-150 77593-474 77672-032
Methyl-2,4-dichloronicotinate 97%
Methyl-2,4-dichloronicotinate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.022 g/mol
MDL Number: MFCD11100222
CAS Number: 442903-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR