3,9-Dibromoperylene 97% (mixture of isomers)

Supplier: Ambeed
Warning

A228963-100MG A228963-1G A228963-250MG A228963-5G
77013-472EA 39.26 USD
77013-472 77013-474 77013-476 77550-478
3,9-Dibromoperylene 97% (mixture of isomers)
3,9-Dibromoperylene

Precaución: for Research Use Only

Formula: C₂₀H₁₀Br₂
MW: 410.10 g/mol
MDL Number: MFCD16658916
CAS Number: 56752-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR