3-Chloro-4-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A228933-10G A228933-1G A228933-250MG
77013-466EA 283 USD
77013-466 77013-468 77013-470
3-Chloro-4-nitrobenzonitrile 98%
3-Chloro-4-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClN₂O₂
MDL Number: MFCD02317167
CAS Number: 34662-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR