2-(Boc-amino)-5-bromo-3-hydroxypyridine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (5-bromo-3-hydroxypyridin-2-yl)carbamate

A228930-10G A228930-1G
77013-460EA 12.74 USD
77013-460 77013-462 77013-464
2-(Boc-amino)-5-bromo-3-hydroxypyridine 97%
2-(Boc-amino)-5-bromo-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O₃
MW: 289.126 g/mol
MDL Number: MFCD12756114
CAS Number: 1207175-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR