3-Bromo-5-hydroxy-2-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-6-methylpyridin-3-ol

A228671-100MG A228671-250MG A228671-1G
76987-244EA 21.22 USD
76987-244 76987-246 77612-222
3-Bromo-5-hydroxy-2-picoline 98%
3-Bromo-5-hydroxy-2-picoline

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.022 g/mol
MDL Number: MFCD09839298
CAS Number: 186593-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR