DL-Homoserine 95%

Supplier: Ambeed
Warning

A228628-100G A228628-10G A228628-1G A228628-250MG
77000-128EA 179 USD
77000-128 77000-130 77000-132 77000-134
DL-Homoserine 95%
DL-Homoserine

Precaución: for Research Use Only

Formula: C₄H₉NO₃
MDL Number: MFCD00002618
CAS Number: 1927-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR