3-Nitro-1H-indole 98%

Supplier: Ambeed
Warning

A228499-10G A228499-1G A228499-250MG A228499-5G
76987-236EA 385 USD
76987-236 76987-238 76987-240 76987-242
3-Nitro-1H-indole 98%
3-Nitro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD13178651
CAS Number: 4770-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR