Methyl 2-(1H-indazol-3-yl)acetate 97%

Supplier: Ambeed
Warning

A228421-100MG A228421-1G A228421-250MG
76987-230EA 43.5 USD
76987-230 76987-232 76987-234
Methyl 2-(1H-indazol-3-yl)acetate 97%
Methyl 2-(1H-indazol-3-yl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
MDL Number: MFCD20527514
CAS Number: 131666-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR