1-Trityl-1H-1,2,4-triazole 97%

Supplier: Ambeed
Warning

A228405-250MG A228405-1G
77013-454EA 6 USD
77013-454 77890-340
1-Trityl-1H-1,2,4-triazole 97%
1-Trityl-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₂₁H₁₇N₃
MDL Number: MFCD00962178
CAS Number: 31250-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR