JIB-04 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
(E)-5-Chloro-2-(2-(phenyl(pyridin-2-yl)methylene)hydrazinyl)pyridine
A228395-1MG
A228395-100MG
A228395-1G
A228395-5MG
A228395-10MG
A228395-250MG
77013-452EA
8.49
USD
77013-452
77600-824
77612-212
77612-214
77683-663
77683-664
JIB-04 98%, Ambeed.Inc
JIB-04
Precaución: For research use only.
Formula:
C₁₇H₁₃ClN₄ MW: 308.76 g/mol |
MDL Number:
MFCD26936344 CAS Number: 199596-05-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...