Toluidine Red 97%

Supplier: Ambeed
Danger

Synonyms: Pigment red 3, 1-(4-Methyl-2-nitrophenylazo)-2-naphthol, 1-((4-Methyl-2-nitrophenyl)diazenyl)naphthalen-2-ol

A228221-1G A228221-5G
77030-978EA 12 USD
77030-978 77872-163
Toluidine Red 97%
Toluidine Red

Precaución: for Research Use Only

Formula: C₁₇H₁₃N₃O₃
MDL Number: MFCD00003910
CAS Number: 2425-85-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR