6-Oxa-1-azaspiro[3.3]heptane hemioxalate 97%

Supplier: Ambeed
Warning

A228167-100MG A228167-1G A228167-250MG A228167-5G
76987-218EA 51 USD
76987-218 76987-220 76987-222 76987-224
6-Oxa-1-azaspiro[3.3]heptane hemioxalate 97%
6-Oxa-1-azaspiro[3.3]heptane hemioxalate

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₆
MDL Number: MFCD22373634
CAS Number: 1380571-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR