L-Ornithine L-Aspartate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2,5-Diaminopentanoic acid (S)-2-aminosuccinic acid salt, H-Orn-OH H-Asp-OH salt

A228049-100G A228049-25G A228049-1000G A228049-500G
77000-110EA 21.22 USD
77000-110 77600-822 77672-014 77672-016
L-Ornithine L-Aspartate 97%
L-Ornithine L-Aspartate

Precaución: for Research Use Only

Formula: C₉H₁₉N₃O₆
MW: 265.27 g/mol
MDL Number: MFCD00058084
CAS Number: 3230-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR