(S)-Ethyl 2-acetamido-3-(4-hydroxy-3,5-diiodophenyl)propanoate 95+%

Supplier: Ambeed
Warning

A227856-1G A227856-25G A227856-5G
77000-104EA 21 USD
77000-104 77000-106 77000-108
(S)-Ethyl 2-acetamido-3-(4-hydroxy-3,5-diiodophenyl)propanoate 95+%
(S)-Ethyl 2-acetamido-3-(4-hydroxy-3,5-diiodophenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₅I₂NO₄
MW: 503.07 g/mol
MDL Number: MFCD00136808
CAS Number: 21959-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR