Apremilast 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-[1-(3-Ethoxy-4-methoxyphenyl)-2-methylsulphonylethyl]-4-acetylaminoisoindoline-1,3-dione

A227494-10MG A227494-1G A227494-1MG A227494-25MG A227494-2MG A227494-50MG A227494-5MG A227494-25G A227494-5G
77013-420EA 10.61 USD
77013-420 77013-422 77013-424 77013-426 77013-428 77013-430 77013-432 77568-208 77683-659
Apremilast 98%
Apremilast

Precaución: For research use only.

Formula: C₂₂H₂₄N₂O₇S
MW: 460.5 g/mol
MDL Number: MFCD18782607
CAS Number: 608141-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR