Ethyl 6-bromo-1H-indazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A227457-100MG A227457-1G A227457-250MG A227457-5G
77030-962EA 47.75 USD
77030-962 77030-964 77030-966 77672-006
Ethyl 6-bromo-1H-indazole-3-carboxylate 97%
Ethyl 6-bromo-1H-indazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₂
MW: 269.10 g/mol
MDL Number: MFCD05664002
CAS Number: 885272-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR