(R)-(+)-3-Hydroxypiperidine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-3-Piperidinol hydrochloride, (3R)-piperidin-3-ol hydrochloride, (R)-Piperidin-3-ol hydrochloride, (R)-3-Hydroxypiperidine hydrochloride
A226941-100G
A226941-10G
A226941-1G
A226941-250MG
A226941-25G
A226941-5G
76987-184EA
197.33
USD
76987-184
76987-186
76987-188
76987-190
76987-192
76987-194
(R)-(+)-3-Hydroxypiperidine hydrochloride 98%
(R)-(+)-3-Hydroxypiperidine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO MW: 137.607 g/mol |
MDL Number:
MFCD00192221 CAS Number: 198976-43-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...