3,5,6-Trichloropicolinic acid 97%

Supplier: Ambeed
Warning

A226913-1G A226913-250MG A226913-5G
77030-956EA 102.91 USD
77030-956 77030-958 77030-960
3,5,6-Trichloropicolinic acid 97%
3,5,6-Trichloropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₂Cl₃NO₂
MW: 226.4446 g/mol
MDL Number: MFCD01086756
CAS Number: 40360-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR