3-Aminocrotononitrile 98%

Supplier: Ambeed
Danger

Synonyms: 3-Iminobutyronitrile, 3-Amino-2-butenenitrile, Diacetonitrile, 3-Aminobut-2-enenitrile

A226867-25G A226867-100G
77000-090EA 8.49 USD
77000-090 77600-820
3-Aminocrotononitrile 98%
3-Aminocrotononitrile

Precaución: for Research Use Only

Formula: C₄H₆N₂
MW: 82.11 g/mol
MDL Number: MFCD00008071
CAS Number: 1118-61-2
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR