Ethyl 5-methylnicotinate 96%

Supplier: Ambeed
Warning

A226731-1G A226731-5G A226731-25G
77030-952EA 11 USD
77030-952 77030-954 77872-157
Ethyl 5-methylnicotinate 96%
Ethyl 5-methylnicotinate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MDL Number: MFCD09030156
CAS Number: 20826-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR