3-(4-Chloro-3-fluorophenyl)propionic acid 97%

Supplier: Ambeed
Warning

A226665-10G A226665-1G
77030-948EA 194.15 USD
77030-948 77030-950
3-(4-Chloro-3-fluorophenyl)propionic acid 97%
3-(4-Chloro-3-fluorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
MDL Number: MFCD04116058
CAS Number: 881189-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR