(3-Chloropyridin-2-yl)methanamine 97%

Supplier: Ambeed
Warning

A226598-100MG A226598-1G A226598-5G
77000-084EA 70.02 USD
77000-084 77000-086 77000-088
(3-Chloropyridin-2-yl)methanamine 97%
(3-Chloropyridin-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
MDL Number: MFCD05664032
CAS Number: 500305-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR