(2E)-3-(5-Methyl-2-furyl)acryloyl chloride 95%

Supplier: Ambeed
Danger

A226262-1G A226262-25G A226262-5G
77000-078EA 12.74 USD
77000-078 77000-080 77000-082
(2E)-3-(5-Methyl-2-furyl)acryloyl chloride 95%
(2E)-3-(5-Methyl-2-furyl)acryloyl chloride

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MDL Number: MFCD12198075
CAS Number: 111252-41-6
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR