L-Thr(Me)-OH 98%

Supplier: Ambeed
Warning

Synonyms: O-Methyl-L-Threonine, H-Thr(Me)-OH, (2S,3R)-2-Amino-3-methoxybutanoic acid

A226243-100MG A226243-10G A226243-1G A226243-250MG A226243-25G A226243-5G
76987-170EA 6.37 USD
76987-170 76987-172 76987-174 76987-176 76987-178 77612-206
L-Thr(Me)-OH 98%
L-Thr(Me)-OH

Precaución: for Research Use Only

Formula: C₅H₁₁NO₃
MW: 133.15 g/mol
MDL Number: MFCD00037767
CAS Number: 4144-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR