2-(4-Piperidyl)-2-propanol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Piperidyl)isopropyl Alcohol

A226204-10G A226204-5G
77000-074EA 118.83 USD
77000-074 77000-076
2-(4-Piperidyl)-2-propanol 98%
2-(4-Piperidyl)-2-propanol

Precaución: for Research Use Only

Formula: C₈H₁₇NO
MW: 143.23 g/mol
MDL Number: MFCD06797159
CAS Number: 22990-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR