4,7-Dibromo-5,6-bis(dodecyloxy)-2,1,3-benzothiadiazole 95%

Supplier: Ambeed
Warning

Synonyms: 4,7-Dibromo-5,6-bis(dodecyloxy)benzo[c][1,2,5]thiadiazole

A226041-100MG A226041-250MG
77013-406EA 41.38 USD
77013-406 77013-408
4,7-Dibromo-5,6-bis(dodecyloxy)-2,1,3-benzothiadiazole 95%
4,7-Dibromo-5,6-bis(dodecyloxy)-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₃₀H₅₀Br₂N₂O₂S
MW: 662.61 g/mol
MDL Number: MFCD27923049
CAS Number: 1313876-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR