(4-Chloro-3,5-dimethylpyridin-2-yl)methanol 95%

Supplier: Ambeed
Warning

A225732-100MG A225732-1G A225732-250MG
77013-394EA 31 USD
77013-394 77013-396 77013-398
(4-Chloro-3,5-dimethylpyridin-2-yl)methanol 95%
(4-Chloro-3,5-dimethylpyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MDL Number: MFCD09859804
CAS Number: 150054-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR