Ethyl 4,5-bis(2-methoxyethoxy)-2-nitrobenzoate 98%

Supplier: Ambeed
Warning

A225612-1G A225612-25G
77030-936EA 27.59 USD
77030-936 77030-938
Ethyl 4,5-bis(2-methoxyethoxy)-2-nitrobenzoate 98%
Ethyl 4,5-bis(2-methoxyethoxy)-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₈
MW: 343.32 g/mol
MDL Number: MFCD06797662
CAS Number: 179688-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR