5-Bromo-3-carboxy-2-fluorobenzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Borono-5-bromo-2-fluorobenzoic acid, 5-Bromo-3-carboxy-2-fluorophenylboronic acid

A225584-1G A225584-250MG A225584-5G
77000-056EA 38.2 USD
77000-056 77000-058 77000-060
5-Bromo-3-carboxy-2-fluorobenzeneboronic acid 97%
5-Bromo-3-carboxy-2-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BBrFO₄
MW: 262.826 g/mol
MDL Number: MFCD09800869
CAS Number: 957120-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR