(1H-Pyrazol-4-yl)methanamine dihydrochloride 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A225201-100MG A225201-10G A225201-1G A225201-250MG A225201-5G
76987-154EA 63.66 USD
76987-154 76987-156 76987-158 77568-178 77683-654
(1H-Pyrazol-4-yl)methanamine dihydrochloride 97%, Ambeed.Inc
(1H-Pyrazol-4-yl)methanamine dihydrochloride

Precaución: For research use only.

Formula: C₄H₉Cl₂N₃
MW: 170.04 g/mol
MDL Number: MFCD09971517
CAS Number: 1172862-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR