4-Chloro-4'-hydroxybenzophenone 97%

Supplier: Ambeed
Warning

Synonyms: (4-Chlorophenyl)(4-hydroxyphenyl)methanone

A225001-1000G A225001-100G A225001-25G A225001-500G
77030-914EA 97 USD
77030-914 77030-916 77030-918 77030-920
4-Chloro-4'-hydroxybenzophenone 97%
4-Chloro-4'-hydroxybenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₉ClO₂
MDL Number: MFCD00002357
CAS Number: 42019-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR