4-(2-Carboxyethyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanoic acid

A224907-1G A224907-250MG A224907-5G
77013-374EA 48.81 USD
77013-374 77013-376 77013-378
4-(2-Carboxyethyl)phenylboronic acid pinacol ester 98%
4-(2-Carboxyethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₄
MW: 276.1400 g/mol
MDL Number: MFCD11053827
CAS Number: 797755-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR