7-Chloro-2-hydroxyquinoline 95%

Supplier: Ambeed
Warning

A224886-100MG A224886-10G A224886-1G A224886-250MG A224886-5G
77000-026EA 54.11 USD
77000-026 77000-028 77000-030 77000-032 77000-034
7-Chloro-2-hydroxyquinoline 95%
7-Chloro-2-hydroxyquinoline

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MW: 179.6030 g/mol
MDL Number: MFCD08460109
CAS Number: 22614-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR