N-tert-Butylcarbamoyl-L-tert-leucine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(3-(tert-Butyl)ureido)-3,3-dimethylbutanoic acid, (S)-2-[3-(tert-Butyl)ureido]-3,3-dimethylbutyric acid

A224873-1G A224873-25G A224873-5G
77013-368EA 12.74 USD
77013-368 77013-370 77013-372
N-tert-Butylcarbamoyl-L-tert-leucine 98%
N-tert-Butylcarbamoyl-L-tert-leucine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₃
MW: 230.31 g/mol
MDL Number: MFCD12796012
CAS Number: 101968-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR