4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))tribenzoic acid 98%

Supplier: Ambeed
Warning

A224838-100MG A224838-1G A224838-250MG A224838-5G
77013-360EA 56.23 USD
77013-360 77013-362 77013-364 77013-366
4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))tribenzoic acid 98%
4,4',4''-(Benzene-1,3,5-triyltris(ethyne-2,1-diyl))tribenzoic acid

Precaución: for Research Use Only

Formula: C₃₃H₁₈O₆
MW: 510.49 g/mol
MDL Number: MFCD17926334
CAS Number: 205383-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR