1,5-Dibromo-2,6-dihydroxynaphthalene 97%

Supplier: Ambeed
Warning

A224759-1G A224759-250MG A224759-25G A224759-5G
77030-906EA 34 USD
77030-906 77030-908 77030-910 77030-912
1,5-Dibromo-2,6-dihydroxynaphthalene 97%
1,5-Dibromo-2,6-dihydroxynaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂O₂
MDL Number: MFCD16251539
CAS Number: 132178-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR