(3-Methylpyridin-2-yl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A224732-1G A224732-250MG
76987-132EA 47 USD
76987-132 76987-134
(3-Methylpyridin-2-yl)methanamine hydrochloride 95%
(3-Methylpyridin-2-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂
MDL Number: MFCD18384218
CAS Number: 153936-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR